Learn More
Fisherbrand™ Plug Without Cable Electrode
This durable coaxial cable for S7 or S8 head electrodes is ideal for connecting screw terminals of pH regulators.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.06000kg
Description
- Plug for connection to screw terminals of pH regulators
- For combined electrodes
- Plug without cable electrode
- S7 cable unplug 5M
Specifications
S7 | |
Plug Without Cable Electrode | |
Plug | |
5 m |