Learn More
Thermo Scientific™ Plumbing Adapter Kit for VersaCool™ Refrigerated Circulating Bath
Plumbing Adapter Kit to be used with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Brand: Thermo Scientific™ 230000006
Additional Details : Weight : 2.00000kg
Description
Utilize the plumbing adapter kit to easily circulate the fluid from the bath to your application. Optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
A complete selection of hose barbs and threaded adapters are included as standard.- 0.25 in. (8mm), 0.5 in (12mm) 8mm and 12mm hose barbs
- 0.25 in. MNPT and 0.5 in. MNPT – ideal for use with quick disconnects
Specifications
Plumbing Adapter Kit | |
Thermo Scientific VersaCool Refrigerated Circulating Bath | |
Plumbing Adapter Kit |