missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

Brand:  Thermo Scientific™ 36500500

 View more versions of this product

Product Code. 10310975

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5uL volume; 31 gauge; 75mm length
GC Autosampler Syringe, Plunger-in-Needle
0.5 μL
Cone
31 G
Reusable
75 mm
TriPlus, AS3000, AS2000, AS200, and AS800 Autosampler
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.