Learn More
Molecular Probes™ Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit
Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit
Brand: Molecular Probes™ A24881
Description
The Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit enables optimal image-based analysis of four key markers of human pluripotent stem cells (PSCs): OCT4, SOX2, SSEA4, and TRA-1-60. This high performance immunocytochemistry (ICC) kit includes a complete set of primary and secondary antibodies, a nuclear DNA stain, and premade buffers for an optimized staining experiment. To characterize pluripotent stem cells using only two markers, we recommend us of the Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) or the Pluripotent Stem Cell Immunocytochemistry Kit (SOX2, TRA-1-60).With the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit, you can:
Confirm Expression of Key PSC Markers
The antibodies included in the kit have been carefully selected and validated for high performance in the ICC analysis of PSCs, whether cultured under feeder-dependent or feeder-free media systems. OCT4 and SOX2 are critical nuclear markers, while SSEA4 and TRA-1-60 are important cell surface markers.
Get More Information Per Sample
The antibody combinations in the kit have been designed to allow for the specific and simultaneous assessment of two markers at a time (e.g., OCT4 & SSEA4 and SOX2 & TRA-1-60) along with nuclear DNA staining to help save time and precious sample.
Generate Beautiful Multiplexed Images
Powered by the superior performance of bright, photostable Alexa Fluor™ dyes, this kit enables the acquisition of beautiful multiplexed images of PSC marker expression using commonly available blue, green, and orange/red filter sets.
Minimize Unnecessary Wash Steps
The streamlined method recommended for this kit eliminates unnecessary wash steps found in standard ICC protocols to help save time and decrease the likelihood of cell loss during the staining protocol, especially when using glass coverslips or chamber slides. The only washes required are after incubation with primary and secondary antibodies.
Process Samples with Confidence
An optimized set of premade fixation, permeabilization, blocking, and wash buffers allows you to focus on answering scientific questions rather than spending time preparing reagents and determining compatibility.
What You Get
Easily scaled to any culture plate, dish, or slide format, the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit comes with reagents sufficient to stain 40 samples using a 50 uL staining volume. (See kit content details below.)

Specifications
2 Color, Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594 | |
40 tests | |
Store at -20°C to 4°C: Store at -20°C to RT: |
|
Immunocytochemistry Kit |
Safety and Handling
- Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Paraformaldehyde
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.