missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEL Monoclonal Antibody (HMB45), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513232
This item is not returnable.
View return policy
Description
MA5-13232 targets Melanoma (gp100) in IHC (P) and WB applications and shows reactivity with Human samples. The MA5-13232 immunogen is extract of pigmented melanoma metastases from lymph nodes.
Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.Specifications
PMEL | |
Monoclonal | |
0.2 mg/mL | |
10mM PBS with 0.05% BSA and 0.05% sodium azide | |
P40967 | |
PMEL | |
Extract of pigmented melanoma metastases from lymph nodes | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
HMB45 | |
Unconjugated | |
PMEL | |
95 kDa melanocyte-specific secreted glycoprotein; 95kDa melanocyte-specific secreted glycoprotein; D10H12S53E; D12S53E; D12S53Eh; gp100; gp87; M-alpha; M-beta; ME20; ME20M; ME20-M; ME20-S; Melanocyte lineage specific antigen GP100; melanocyte protein mel 17; melanocyte protein PMEL; Melanocyte protein Pmel 17; melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; P26; PMEL; Pmel 17; PMEL17; premelanosome protein; Secreted melanoma-associated ME20 antigen; SI; SIL; SILV; silver homolog; silver locus protein; silver locus protein homolog; silver, mouse, homolog of | |
Mouse | |
Protein A/G | |
RUO | |
6490 | |
4° C | |
Liquid |