missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ PHMT Thermoshaker for Microtubes and Microplates
Accelerates in 3 seconds for fast start but also operates smoothly and quietly, this Grant Instruments™ PMHT Thermoshaker for Microtubes and Microplates is ideal for DNA extraction, enzyme reactions and cellular biology techniques.
£1600.60 - £2146.50
Specifications
Orbit | 2 mm |
---|---|
Speed Range | 250 to 1400 rpm (increments 10 rpm) |
Temperature Range (Metric) | Ambient +5° to 100°C |
Temperature Setting | 5°C to 100°C |
Temperature Uniformity | 0.1°C at 37°C; 0.2°C at 60°C; 0.2°C at 100°C |
Product Code | Brand | Holds | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Model | Price | Quantity & Availability | ||||
15201176
|
Grant Instruments™
PHMT-PSC96 |
96-well PCR plate | PHMT-PSC96 |
£1600.60
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
13479429
|
Grant Instruments™
PHMT-PSC24 |
24 x 2 mL Microtubes | PHMT-PSC24 |
£1860.30
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
15163706
|
Grant Instruments™
PHMT-PSC24N |
24 x 1.5 mL Microtubes | PHMT-PSC24N |
£2146.50
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||||
15291166
|
Grant Instruments™
PHMT-PSC32 |
32 x 0.2 mL and 12 x 1.5 mL Microtubes | PHMT-PSC32 |
£1600.60
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
11411048
|
Grant Instruments™
PHMT-PSC18 |
32 x 0.5 mL/1.5 mL Microtubes | PMHT-PSC18 |
£1600.60
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
Description
- Combines three instruments in one:
- Microtube and microplate thermoshaker
- Compact benchtop incubator
- Microtube and microplate shaker in cold or ambient temperatures
- 2-line LCD display clearly indicates both set and actual values for temperature, shaking speed and time.
- Interchangeable block for 20 or 12 microtubes provides flexibility for an easy change in application.
- Operates extremely smoothly, quietly and consistently.
- Includes ‘soft’ start function for delicate samples.
- Fast start - acceleration 3 seconds.
- Compact and sturdy, with a low profile and small footprint.
Specifications
2 mm | |
Ambient +5° to 100°C | |
0.1°C at 37°C; 0.2°C at 60°C; 0.2°C at 100°C | |
130 mm | |
4 kg |
250 to 1400 rpm (increments 10 rpm) | |
5°C to 100°C | |
230 mm | |
205 mm |