missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PMP Class A Measuring Flask

PMP flask with polypropylene stopper

Brand:  BRAND™ 36182

23.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10766833

  • £34.95 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Error limits according to class A, DIN EN ISO 1042
  • Includes one batch certificate
  • Calibrated to contain (TC, In)
  • Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    1000 mL
    PP stopper
    0.4 mL
    A
    PMP
    Ground joint: NS 24/29
    Autoclavable
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers