missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PMP Griffin Beaker

Product Code. 12723409
Click to view available options
Capacity (Metric):
25 mL
100 mL
150 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 12
Pack of 6
This item is not returnable. View return policy

Product Code. 12723409

Brand: VITLAB™ 61803

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Make your job easier and safer while handling a wide range of liquids with these polymethylpentene beakers. VITLAB™ PMP Griffin Beakers feature an easy to read, embossed red scale.

  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C

Specifications

Color Crystal Clear
Capacity (Metric) 500 mL
Material PMP
For Use With (Application) For Volume Measurement
Height (Metric) 117.5 mm
Type Griffin Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Red Printed Scale
Style Griffin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.