missing translation for 'onlineSavingsMsg'
Learn More
PMPA (NAALADase inhibitor), Tocris Bioscience™ Product Code.: 18044602

PMPA (NAALADase inhibitor), Tocris Bioscience™

Product Code. 18044602
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18044602

Brand: Tocris Bioscience 1380/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GCP II inhibitor

Highly potent and selective inhibitor of glutamate carboxypeptidase 2 (GCP II/N-acetylated α-linked dipeptidase/NAALADase) with a Ki value of 275 pM. Neuroprotective; protects against glutamate-mediated motor neuron degeneration and reduces volume of injury following middle cerebral artery occlusion (MCAO).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173039-10-6
Molecular Formula C6H11O7P
Molecular Weight (g/mol) 226.121
InChI Key ISEYJGQFXSTPMQ-UHFFFAOYSA-N
Synonym 2-phosphonomethyl pentanedioic acid, 2-pmpa, pmpa naaladase inhibitor, 2-phosphonomethyl-pentanedioic acid, chembl47009, pmpa naaladaseinhibitor, d0f3zo, d0y6ub, 2-pmpa naaladaseinhibitor
PubChem CID 10130754
IUPAC Name 2-(phosphonomethyl)pentanedioic acid
SMILES C(CC(=O)O)C(CP(=O)(O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 226.12
Percent Purity >98%
Chemical Name or Material PMPA (NAALADase inhibitor)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PMPA (NAALADase inhibitor), Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.