missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Pocket Turbidimeter Turb 430 T / Set
Three-point calibration automatically with user guidance and calibration and documentation
Brand: WTW™ 600326
2340.37 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The turbidity meters Turb™ 430 IR acc. to DIN ISO 27027 and Turb™ 430 T acc. to US EPA 180.1 meet the requirements of the nephelometric measurement at 90° stray light for drinking water with highest precision, especially in the range < 1 NTU. The measuring range comprises 0-1100 NTU/FNU with automatic measuring range switching and is therefore suitable for water analysis as well as quality control, such as gasoline, cell cultures, as well as in the food and beverage industries.The measurement and automatic 3-point calibration runs self-explanatory and menu-controlled. The calibration takes place via an AMCO Clear™ standard set (0.02-10-1000 NTU). Up to 1000 data sets including sample ID numbers can be stored and later put out GLP-compliant via the PC software LSdata . The quality assurance of the measured results is supported by calibration interval setting with documentation.- Manufacturing tolerance of only ±1% for highest result accuracy
- Long-term stable particle distribution and size
- No health hazard
- Easy to dispose of
- N.I.S.T. traceability

Specifications
Pocket Turbidity Meter Turb 430 T / Set | |
Infrared LED | |
Universal Power Supply RB Flex |
White Light or Infrared | |
Drinking / measurements <1 NTU> - 3-point calibration automatically with user guidance and calibration and documentation | |
US EPA |
Product Suggestions
Customers who viewed this item also viewed
CE_10789303.pdf
File size: 505.5 kB
CE Certificate
File size: 505.5 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Pocket Turbidimeter Turb 430 T / Set > For Use With: Drinking / measurements <1 NTU> - 3-point calibration automatically with user guidance and calibration and documentation
Spot an opportunity for improvement?Share a Content Correction