Learn More
Thermo Scientific™ PocketTips™ Pipette Tips
Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.
£186.41 - £3904.07
Specifications
Filtered | Non-filtered |
---|---|
Sterility | Non-sterile |
Color | Clear |
Product Line | PocketTip |
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
12997207
|
Thermo Scientific™
FLIPR384B-250 |
250 nL | Molecular Devices FLIPR 384-well Head | N/A | |||||
12907217
|
Thermo Scientific™
FLIPR384B-50 |
50 nL | Molecular Devices FLIPR 384-well Head | N/A | |||||
12937217
|
Thermo Scientific™
FX384P30-50 |
50 nL | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384 | N/A | |||||
12947217
|
Thermo Scientific™
FX96P20-100 |
100 nL | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96 | N/A | |||||
12967217
|
Thermo Scientific™
FX96P20-50 |
50 nL | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96 | N/A | |||||
12977217
|
Thermo Scientific™
FX96P20-500 |
500 nL | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96 | N/A | |||||
11662020
|
Thermo Scientific™
PE384P30-100 |
100 nL | Packard 384-well, PerkinElmer | N/A | |||||
11672020
|
Thermo Scientific™
PE384P30-50 |
50 nL | Packard 384-well, PerkinElmer | N/A | |||||
11682020
|
Thermo Scientific™
PE96P235-100 |
100 nL | Packard 96-well, PerkinElmer | N/A | |||||
11692020
|
Thermo Scientific™
PE96P235-250 |
250 nL | Packard 96-well, PerkinElmer | N/A | |||||
11902221
|
Thermo Scientific™
STAR384P30-50 |
50 nL | Hamilton MICROLAB STAR | N/A | |||||
Description
Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.
Features:
- Transfer samples directly to the assay plate
- Combine sample and assay material with soft in-tip mixing
- Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
- Color: Clear
Recommended for:
Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
Non-filtered | |
Non-sterile | |
Clear | |
PocketTip |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.