missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL2 Monoclonal Antibody (211816), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523932
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
PODXL2 Monoclonal antibody specifically detects PODXL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
PODXL2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NZ53 | |
PODXL2 | |
Mouse myeloma cell line NS0-derived recombinant human Endoglycan/PODXL2 Gly33-Thr500 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry | |
211816 | |
Unconjugated | |
PODXL2 | |
D130074J02Rik; EG; endoglycan; LOW QUALITY PROTEIN: podocalyxin-like protein 2; PODLX2; podocalyxin like 2; podocalyxin-like 2; podocalyxin-like protein 2; Podxl2; RGD1305815; sialomucin protein; UNQ1861/PRO3742 | |
Mouse | |
Protein A/G | |
RUO | |
50512 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |