missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polar Silica-Based SPE Columns, Silica (SI), Medical-grade polypropylene with PE frits, 500 mg/25 mL, Honeywell Fluka
Click to view available options
Quantity:
100 Pc.
Packaging:
Fiberboard Box
Description
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 Pc. |
Packaging | Fiberboard Box |
Grade | Analytical |
Chemical Name or Material | Polar Silica-Based SPE Columns, Silica (SI), Medical-grade polypropylene with PE frits, 500 mg/25 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'