Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter Set for LED Ring Light
Polarization Filter Set for LED 66 mm ID Ring Light. Includes polarizer and analyzer. For use with VisLED brightfield Ring Light, EasyLED Ring Light and EasyLED Ring Light Plus.
Brand: SCHOTT AG Lighting and Imaging™
Code : A0
Additional Details : Weight : 0.20000kg
Description
- Polarization system for brightfield ringlights.
- Turnable polarizer with adapting thread and sliding filter as analyzer.
- Provides polarized light by a polarizing and analyzing filter within the beam path.
- Can be used very efficiently in Stereo Microscopy to contrast some inner structure of material or to identify metal particles within others.
Specifications
Polarization Filter | |
Stereo Microscopy Illumination | |
Polarizer and Analyzer |
66 mm | |
EasyLED Ringlights or VisiLED Ringlight S80-55 |