Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter for Spotlight
Turnable Polarization Filter with adapting thread for use with EasyLED single or double Spotlight Plus. Available in Red, Blue, Green, Yellow or Halogen and a polarization filter.
Brand: SCHOTT AG Lighting and Imaging™ 600600
Additional Details : Weight : 5.00000kg
Description
- Color filters for EasyLED single or double Spotlight Plus.
- Use for fixing at spot light with thread.
- Available colors: red, blue, green, yellow and halogen
- Polarization filter available.
Specifications
Polarization Filter | |
Stereo Microscopy Illumination | |
M22.5 x 0.5 |
Polarized light makes properties of the materials visible (polymeric structures) | |
EasyLED Spotlight Plus (Single or Double) |