Learn More
Thermosafe™ PolarPack™ Refrigerant Gel Packs
Economical and practical refrigerant pack choice for shipping temperature-sensitive material such as food products, pharmaceuticals, and more. Thermosafe™ PolarPack™ Refrigerant Gel Packs are enclosed in durable polyethylene film.
Brand: Thermosafe™
Code : 04
Additional Details : Weight : 15.15600kg
Description
- Reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products.
- Multi-layered exterior minimizes surface condensation which protects product from moisture damage.
- Outer film layer assures products arrive in top condition by absorbing condensation.
- Phase change formulas ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2.
- Laminated, metalized film exterior provides radiant heat barrier, resists punctures.
- Available in several sizes
- Reusable
- Non-toxic
Specifications
72 Pk./Cs. | |
8 oz./Pack | |
6.5 x 3.5 x 0.75 in. |