Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Confidently store and monitor temperature-sensitive samples
Brand: Cole-Parmer™ 44260-71
This item is not returnable.
View return policy
Description
- Improve sample protection by maintaining specific temperature ranges for extended time periods
- Maintain sample integrity with Traceable™ temperature monitoring
- Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
95 L | |
78.74 cm | |
33.5 in. | |
85.09 cm |
31 in. | |
8.5 in. | |
21.6 cm | |
Aluminum |