Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Confidently store and monitor temperature-sensitive samples
Brand:  Cole-Parmer™ 44260-72
 View more versions of this product
Product Code. 16612874
  • £7120.00 / Each
Explore more special offers
Description
Description
  • Improve sample protection by maintaining specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable™ temperature monitoring
  • Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Specifications
115 L
88.27 cm
33.5 in.
89.09 cm
34.75 in.
8.5 in.
21.6 cm
Aluminum