Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Confidently store and monitor temperature-sensitive samples
Brand:  Cole-Parmer™ 44260-75
 View more versions of this product
Product Code. 16642874
  • £2280.00 / Each
Explore more special offers
Description
Description
  • Improve sample protection by maintaining specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable™ temperature monitoring
  • Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Specifications
47 L
71.76 cm
21.3 in.
54.10 cm
28.25 in.
5 in.
12.7 cm
Aluminum