Learn More
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank, with self-pressurized liquid nitrogen, makes LN2 discharge safe and simple
£2705.00 - £3603.81
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16639561
|
Cole-Parmer™
4426068 |
30 L |
£2705.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
16649561
|
Cole-Parmer™
4426069 |
50 L |
£3603.81
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Description
- Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
- Unique design utilizes pressure from LN2 to discharge LN2 to other containers
- Use as a self-pressurized LN2 backup system for cryogenic freezers
- 5 year vacuum warranty
Specifications
Stainless Steel |