missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poliovirus receptor Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27486-1-AP-20UL
This item is not returnable.
View return policy
Description
PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or alpha and kappa,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbone, respectively. Two other splice variants, beta and gamma encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues. MW of PVR is.Specifications
Poliovirus receptor | |
Polyclonal | |
Unconjugated | |
PVR | |
CD155, HVED, Necl 5, NECL5, Nectin like protein 5, Poliovirus receptor, PVR, PVS, TAGE4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5817 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15151 | |
PVR | |
Poliovirus receptor Fusion Protein Ag26657 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Poliovirus receptor Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction