missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 188, Thermo Scientific Chemicals

Product Code. 15407319
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C5H10O2
Synonym Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer

Specifications

CAS 9003-11-06
Color White
Content And Storage Ambient temperatures
Quantity 1 kg
Solubility Miscible with water.
Form Solid
Chemical Name or Material Poloxamer 188

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.