Learn More
POLR3E Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15022-1-AP-150UL
This item is not returnable.
View return policy
Description
The POLR3E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human POLR3E. This antibody recognizes human, mouse, rat antigen. The POLR3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
POLR3E | |
Polyclonal | |
Unconjugated | |
150 μL | |
55718 | |
IgG |
Western Blot, Immunoprecipitation, ELISA | |
267 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |