missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
2 g
Description
Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) research and devices. It is also used in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Physical Form | Powder/Chunks |
Solubility Information | Soluble (50mg/ml) in chlorinated solvents such as chloroform,chlorobenzene,dichlorobenzene and trichlorobenzene. |
Quantity | 100 mg |
Sensitivity | Light sensitive |
Chemical Name or Material | Poly (3-hexylthiophene-2,5-diyl) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'