missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, Thermo Scientific™

Product Code. 10781841
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color White
CAS 62649-23-4
Physical Form Granules
Molecular Formula (C3H5NO)n
MDL Number MFCD00084393
Synonym 2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate, sodium acrylamide acrylic acid acrylate
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol
Quantity 5 g
InChI Key DTBMFUOWAASNDB-UHFFFAOYSA-M
SMILES NC(=O)C(-*)C-*
IUPAC Name sodium prop-2-enamide prop-2-enoic acid prop-2-enoate
Molecular Weight (g/mol) 237.19
PubChem CID 23696299
Grade Pure
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Chemical Name or Material Poly(acrylamide), carboxyl modified
Show More Show Less
Product Identifier
  • Poly(acrylamide), carboxyl modified

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.