missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White to Yellow |
Physical Form | Granules |
Linear Formula | [-CH2CH(CONH2)-]n |
Density | 1.3000g/mL |
Synonym | acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid |
Quantity | 5 g |
IUPAC Name | prop-2-enamide |
Molecular Weight (g/mol) | 71.08 |
PubChem CID | 6579 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'