missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, Thermo Scientific Chemicals

Product Code. 10675432
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-05-8
Molecular Formula (C3H5NO)n
MDL Number MFCD00084392
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C(-*)C-*

Specifications

Color White to Yellow
Physical Form Granules
Linear Formula [-CH2CH(CONH2)-]n
Density 1.3000g/mL
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid
Quantity 250 g
IUPAC Name prop-2-enoic acid
Molecular Weight (g/mol) 72.06
PubChem CID 6581
ChEBI CHEBI:18308
Formula Weight 5 to 6.000.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 13% max.
Packaging Glass bottle
Specific Gravity 1.3
Chemical Name or Material Poly(acrylamide)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'