missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(Allylamine Hydrochloride), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Poly(allylamine hydrochloride) is used in film and polymer industries and cell encapsulation. Further, it is used to make redox hydrogel- modified electrodes for measuring enzyme responses. In addition, it is used in the preparation of an antireflective coating consists of alternate layers of poly(allyamine hydrochloride) and silica nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Yellow |
Physical Form | Powder |
Solubility Information | Soluble in water. |
Quantity | 10 g |
Chemical Name or Material | Poly (Allylamine Hydrochloride) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'