Learn More
Gibco™ Poly-D-Lysine
Description
Poly-D-Lysine is a chemically synthesized extracellular matrix used to facilitate cell adhesion to tissue culture treated plastic and glass surfaces. Poly-D-Lysine contains polymers with a molecular weight between 50,000 – 150,000 Daltons which makes this product ideal for neuronal culture applications. This ready-to-use product is provided as a liquid at a concentration of 0.1 mg/mL. When used on Nunc cell culture plastics with B-27 Supplements or the B-27 Plus Neuronal Culture System, Poly-D-Lysine has been proven to maintain cell viability and normal growth characteristics in multiple neuronal cell types. Key features include:
- Synthetic and animal origin-free
- Sterile-filtered for cell culture applications
- Validated with B-27 Supplement and B-27 Plus Neuronal Culture System when used with Nunc plastic and glass bottom dishes
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Cell Type | Mammalian Cells |
| Classification | Animal Origin-free |
| Concentration | 0.1 mg/mL |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Extracellular Matrix |
| Sterility | Sterile |
| Product Line | Gibco |
| Purity or Quality Grade | Cell Culture Grade |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.