missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-D-Lysine Hydrobromide, MP Biomedicals
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
Specifications
Specifications
Quantity | 100 mg |
Formula Weight | 4,000 to 15,000 |
Chemical Name or Material | Poly-D-Lysine Hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'