missing translation for 'onlineSavingsMsg'
Learn More

Poly-D-Lysine Hydrobromide, MP Biomedicals

Product Code. 11458350
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 27964-99-4
Molecular Formula C6H15BrN2O2
Molecular Weight (g/mol) 227.102
InChI Key MEXAGTSTSPYCEP-NUBCRITNSA-N
Synonym d-lysine hydrobromide
PubChem CID 87493163
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrobromide
SMILES C(CCN)CC(C(=O)O)N.Br

Specifications

Quantity 500 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-D-Lysine Hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'