missing translation for 'onlineSavingsMsg'
Learn More
Poly(ethylene glycol), average M.W. 200, Thermo Scientific Chemicals Product Code.: 10654702

Poly(ethylene glycol), average M.W. 200, Thermo Scientific Chemicals

Product Code. 10654702
1 L, Plastic bottle
Click to view available options
Quantity:
5 mL
1 L
5 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10654702

Brand: Thermo Scientific Acros 192210010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Liquid
Melting Point -65.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1250g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 1 L
Formula Weight 200
Flash Point 171°C
Hydroxyl Value 534 to 591mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.4580 to 1.4610
Specific Gravity 1.125
Viscosity 60-70 mPa.s (20°C)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465980 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number192210010,192210051
Lot NumberA0459672 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog Number192210010,192210051,192210050
Lot NumberA0455051 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number192210010,192210051,192210050
Lot NumberA0394420 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number192210010,192210051,192210050
Lot NumberA0401150 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number192210010,192210051,192210050
compliance-icons
Product Identifier
  • Poly(ethylene glycol), average M.W. 200
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Poly(ethylene glycol), average M.W. 200, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.