missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 4000, Thermo Scientific Chemicals

Product Code. 10783902
Click to view available options
Quantity:
5 kg
Packaging:
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Flakes
Melting Point 54.0°C to 58.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15,7688
Quantity 5 kg
Formula Weight 4000
Infrared Spectrum Authentic
Packaging Plastic drum
Water 0.5% max.
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less
Product Identifier
  • Poly(ethylene glycol), average M.W. 4000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'