missing translation for 'onlineSavingsMsg'
Learn More
Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals Product Code.: 10471621

Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals

Product Code. 10471621
1 L, Plastic bottle
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10471621

Brand: Thermo Scientific Acros 192240010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Viscous Liquid After Melting
Melting Point 17.0°C to 22.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1265g/mL
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 1 L
Formula Weight 600
Flash Point 252°C
Hydroxyl Value 178 to 197mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.4660 to 1.4700
Specific Gravity 1.1265
Viscosity 16-19 mPa.s (20°C)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 600
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465948 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number192240051,192240010
Lot NumberA0450038 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number192240051,192240010
Lot NumberA0402064 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number192240051,192240010
Lot NumberA0436574 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number192240051,192240010
Lot NumberA0426098 Certificate TypeCertificate of Analysis Date26/01/2021 Catalog Number192240051,192240010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Poly(ethylene glycol), average M.W. 600, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.