missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol), average M.W. 6000, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | White flakes or powder |
Melting Point | 55.0°C to 60.0°C |
Linear Formula | H(OCH2CH2)nOH |
Merck Index | 15, 7688 |
Solubility Information | Solubility in water: 550g/L (20°C). Other solubilities: soluble in thf |
Quantity | 1 kg |
Formula Weight | 6000 |
Hydroxyl Value | 17 to 20mg KOH/g |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'