missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), ∽ M.W. 300,000, Thermo Scientific Chemicals

£22.41 - £291.45

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10523931
View Documents
Thermo Scientific
178600050
5 g Glass bottle
£22.41
5g
In Stock
to see stock.
Add to basket
 
10751071
View Documents
Thermo Scientific Acros
178602500
250 g Glass bottle
£97.38
250g
In Stock
to see stock.
Add to basket
 
10086093
View Documents
Thermo Scientific Acros
178600010
1 kg Plastic bottle
£291.45
1kg
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25322-68-3
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
(C2H4O)n
MFCD01779601
PEO
[H]OCCO

Specifications

White
Powder
(C2H4O)n
MFCD01779601
PEO
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
Authentic
Poly(ethylene oxide), Approximate M.W. 300.000
25322-68-3
65.0°C to 67.0°C
(-CH2CH2O-)n
15, 7688
Solubility in water: soluble. Other solubilities: soluble in many organic solvents
[H]OCCO
62.07
1% max.
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TQ4100000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.