Learn More
Cytiva Poly(I)-Poly(C)
For stimulation of interferon production in vitro. Cytiva Poly(I)-Poly(C) is supplied as a sterile lyophilised powder.
Brand:  Cytiva 27-4732-01
1740.97 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 11545784
  • £2017.00 / 265mg
Explore more special offers
Description
Description
  • Double-stranded polyribonucleotide poly(I)−poly(C) has been shown to induce interferon in rabbit cells
  • This product is dispensed by weight of dry material
  • This weight includes polymer, residual salt and residual water
  • Content of polymer may vary from lot to lot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
265 mg
White
Powder