missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrochloride
Poly-L-Lysine Hydrochloride is a cellular attachment factor in culture applications.
£265.00 - £1175.54
Specifications
CAS | 26124-78-7 |
---|---|
Color | Whitish |
Content And Storage | -20°C |
Format | Powder |
For Use With (Application) | Cellular attachment factor in culture applications |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17348583
|
MP Biomedicals™
0215269090 |
500 mg |
£1175.54
500mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17338583
|
MP Biomedicals™
0215269080 |
100 mg |
£265.00
100mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
- Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
- When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
- Polymers of both D- and L-lysine are used to coat solid surfaces.
- Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
Specifications
26124-78-7 | |
-20°C | |
Cellular attachment factor in culture applications |
Whitish | |
Powder | |
>20,000 g/mol |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.