missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 2.000, Thermo Scientific Chemicals

Product Code. 10489890
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Color White to Gray
Physical Form Crystals or Crystalline Powder
Acid Value 0.1 mg KOH/g max.
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Quantity 5 g
Hydroxyl Value 53 to 58 mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Viscosity 300 cSt (25°C)
Water 0.15% Max.
Chemical Name or Material Poly(propylene glycol), Average M.W. 2.000
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'