missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(sodium-p-styrenesulfonate), average M.W. 70.000, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Tan to White |
Physical Form | Powder |
Linear Formula | [-CH2CH(C6H4SO3Na)-]n |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in lower glycerols |
Quantity | 500 g |
IUPAC Name | 4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Chemical Name or Material | Poly(sodium-p-styrenesulfonate), Average M.W. 70.000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'