missing translation for 'onlineSavingsMsg'
Learn More

Poly(sodium-p-styrenesulfonate), average M.W. 70.000, Thermo Scientific Chemicals

Product Code. 10012830
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Color Tan to White
Physical Form Powder
Linear Formula [-CH2CH(C6H4SO3Na)-]n
Solubility Information Solubility in water: soluble. Other solubilities: soluble in lower glycerols
Quantity 5 g
IUPAC Name 4-ethenylbenzene-1-sulfonic acid
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Poly(sodium-p-styrenesulfonate), Average M.W. 70.000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'