missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals

Product Code. 11389418
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Poly(styrene-divinylbenzene) is used in chromatographic separation medium. It is also used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether and tert-butylcatechol. It is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions. Styrene-divinylbenzene copolymer is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-70-7
Molecular Formula C24H27NO5
Molecular Weight (g/mol) 409.48
MDL Number 9003-70-7
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
Synonym styrene/divinylbenzen, divinylbenzene-styrene, styrene divinylbenzene, styrene-divinylbenzene, styrene/divinylbenzene, styrene-divinyl benzene, benzene, diethenyl-, polymer with ethenylbenzene, brominated, st dvb, divinylbenzene; styrene, 1,2-diethenylbenzene; styrene
PubChem CID 174664
IUPAC Name 1,2-bis(ethenyl)benzene;styrene
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O

Specifications

Physical Form Beads
Solubility Information Insoluble in water.
Quantity 100 g
Mesh Size 100 to 200 Mesh
Odor Odorless
Chemical Name or Material Poly (styrene-divinylbenzene), 1% cross-linked

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.