missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh, Thermo Scientific™

Product Code. 11408590
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89551-24-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00146442
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Physical Form Powder
Quantity 25 g
Mesh Size 200 to 400 Mesh
Chemical Name or Material Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'