missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(tetrafluoroethylene)
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
250g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Linear Formula | (-CF2CF2-)n |
| Density | 2.0000g/mL |
| Quantity | 100 g |
| Infrared Spectrum | Authentic |
| Loss on Drying | 1% max. |
| Packaging | Plastic bottle |
| Chemical Name or Material | Poly(tetrafluoroethylene) |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction