missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(Vinyl Alcohol), Thermo Scientific Chemicals
£31.56 - £333.66
Chemical Identifiers
CAS | 9002-89-5 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 44.05 |
MDL Number | MFCD00081922 |
InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
PubChem CID | 11199 |
IUPAC Name | ethenol |
SMILES | OC(-*)C-* |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10083963
|
Thermo Scientific Acros
396760250 |
25 g | Plastic bottle |
£31.56
25g |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10711244
|
Thermo Scientific Acros
396765000 |
500 g | Plastic bottle |
£93.57
500g |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10276983
|
Thermo Scientific Acros
396760010 |
1 kg | Plastic bottle |
£155.78
1kg |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||||
11966971
|
Thermo Scientific Acros
396760025 |
2.5 kg | Plastic Bottle |
£333.66
2.50kg |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
11199 | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
ethenol |
Specifications
White to Yellow | |
9002-89-5 | |
160.0°C to 240.0°C | |
(C2H4O)n | |
MFCD00081922 | |
Solubility in water: soluble in hot water. | |
OC(-*)C-* | |
44.05 | |
87.0 to 89.0% (degree of hydrolysis) | |
3.5 to 4.5cP (4% aq. solution, 20°C) Brookfield |
4.5 to 6.5 (4% aq. soln.) | |
Powder | |
90%% (Na2O) max. | |
[-CH2CH(OH)-]n | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
11199 | |
Authentic | |
Poly(vinyl alcohol) |
RUO – Research Use Only