Learn More
Poly(vinylidene fluoride), Thermo Scientific Chemicals
£120.00 - £968.00
Chemical Identifiers
CAS | 24937-79-9 |
---|---|
Molecular Formula | (C2H2F2)n |
Molecular Weight (g/mol) | NaN |
MDL Number | MFCD00084470 |
InChI Key | BQCIDUSAKPWEOX-UHFFFAOYSA-N |
Synonym | vinylidene fluoride, 1,1-difluoroethylene, ethene, 1,1-difluoro, vinylidene difluoride, pvdf, genetron 1132a, ethylene, 1,1-difluoro, halocarbon 1132a, poly vinylidene fluoride, kynar |
PubChem CID | 6369 |
ChEBI | CHEBI:82550 |
IUPAC Name | 1,1-difluoroethene |
SMILES | FC(F)(-*)C-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375168
|
Thermo Scientific Alfa Aesar
044080.36 |
500 g |
£120.00
500g |
In Stock
Log in to see stock. |
|||||
11385168
|
Thermo Scientific Alfa Aesar
044080.A3 |
2 kg |
£363.00
2kg |
In Stock
Log in to see stock. |
|||||
11365168
|
Thermo Scientific Alfa Aesar
044080.A9 |
10 kg |
£968.00
10kg |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
Description
Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24937-79-9 | |
NaN | |
BQCIDUSAKPWEOX-UHFFFAOYSA-N | |
6369 | |
1,1-difluoroethene |
(C2H2F2)n | |
MFCD00084470 | |
vinylidene fluoride, 1,1-difluoroethylene, ethene, 1,1-difluoro, vinylidene difluoride, pvdf, genetron 1132a, ethylene, 1,1-difluoro, halocarbon 1132a, poly vinylidene fluoride, kynar | |
CHEBI:82550 | |
FC(F)(-*)C-* |
Specifications
White | |
Powder | |
(C2H2F2)n | |
vinylidene fluoride, 1,1-difluoroethylene, ethene, 1,1-difluoro, vinylidene difluoride, pvdf, genetron 1132a, ethylene, 1,1-difluoro, halocarbon 1132a, poly vinylidene fluoride, kynar | |
BQCIDUSAKPWEOX-UHFFFAOYSA-N | |
1,1-difluoroethene | |
6369 | |
Odorless |
24937-79-9 | |
155°C to 160°C | |
MFCD00084470 | |
Soluble in organic esters,dimethylacetamide and amines. Insoluble in water. | |
FC(F)(-*)C-* | |
NaN | |
CHEBI:82550 | |
Poly (vinylidene fluoride) |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only