Learn More
Buerkle™ Polyacetal Hose Clips
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
Brand:  Buerkle™ 8678-0015
3.72 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10347752
  • £7.95 / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hose clips made of Polyacetal (POM)
  • Featured in varying diameters
  • POM is white, sterilizable, does not rust, permanently elastic and hygienic.
Specifications
Specifications
Hose Clip
Hoses
13 to 15 mm
Polyacetal
10 Pc./Pk.
White