missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Polyacetal Hose Clips
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
Brand: Buerkle™ 8678-0014
3.72 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hose clips made of Polyacetal (POM)
- Featured in varying diameters
- POM is white, sterilizable, does not rust, permanently elastic and hygienic.
Specifications
Hose Clip | |
Hoses | |
12 to 14 mm |
Polyacetal | |
10 Pc./Pk. | |
White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction