missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Polyacetal Joint Clips

Product Code. p-8002682
Click to view available options
Color:
Blue
Brown
Green
Mint
Ochre
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10

Product Code. 11991528

Brand: Quickfit™ KC19

Only null left
Add to basket
Edge
Add to basket

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10/Pk.
Type Joint Clamp
Material Polyacetal
For Use With (Equipment) Glassware with conical joints
Size Joint Size 19/26
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.