Learn More
Palintest™ Polyacrylate Square Tube
For use with Palintest™ comparator
Brand: Palintest™ PT521/5
Code : 04
Additional Details : Weight : 0.04512kg
Description
For use with Palintest™ comparator
The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.Specifications
10 mL | |
5Pack | |
13.5 mm |
Polyacrylate | |
5 Pack | |
Test Tube |